Pro™ Goggle and mask, FilterSpec®
Leverandør: JSP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AGE130-201-100 ASG03B-101-100
JSPAAGE130-201-100EA 896 DKK
JSPAAGE130-201-100 JSPAASG03B-101-100
Pro™ Goggle and mask, FilterSpec®
Beskyttelsesbriller
A unique combination of eye and respiratory protection.
  • Lightweight for user comfort
  • Anti-mist and anti-scratch goggle
  • Disposable filter mask element
  • Supplied with three filters
  • PVC goggle frame, polycarbonate lens
  • Polyethylene filter holder
The unique FilterSpec® Pro™ effortlessly combines both eye and respiratory protection. The 5.5 base, one-piece polycarbonate lens offers optimal coverage and class 1 optics while the ergonomically shaped soft goggle housing provides excellent levels of comfort and unbeatable levels of protection. Anti-Scratch and anti-mist coatings are standard.
The easily replaced filters are made from three layers for exceptional performance. Available with a low breathing resistance valve to reduce moisture and CO₂ build-up. Includes mask holder, three filters, and goggle with clear hardcoat and anti-mist lens.
Certificeringer: Respiratory Protection: EN 149:2001 + A1:2009. Eye Protection: EN 166:2002.