VISTAlens® safety helmet, EVO®
Leverandør: JSP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AMA170-004-F00 AMB170-005-F00 AMB170-006-F00 AMB170-007-F00 AMB177-T04-F00
JSPAAMA170-004-F00EA 640 DKK
JSPAAMA170-004-F00 JSPAAMB170-005-F00 JSPAAMB170-006-F00 JSPAAMB170-007-F00 JSPAAMB177-T04-F00
VISTAlens® safety helmet, EVO®
Hjelme Sikkerhedshjelme
EVO® VISTAlens® is a next generation, feature-rich safety helmet based on the proven Evolution® head protection technology. Incorporating a fully retractable optical class 1 integrated over spec. The eyewear is easy to deploy, adjust and maintain ensuring protection is on hand whenever it is required.
  • Feature-rich safety helmet with class 1 optical integrated eyewear
  • Easy to deploy eye protection
  • Safety cell prevents ricocheting debris injuring the scalp
  • Wear prescription eyewear without compromise
  • Wheel ratchet adjustment
  • ABS shell
  • K&N rated
The EVO® VISTAlens® safety helmet is compact and lightweight. The helmet combines a super strong ABS shell designed for superior all day protection with the comfort benefits of the Evolution® 3D-Adjustment™ harness system. A fully adjustable wheel ratchet and unique 3-D adjustment secures the helmet, while a 6-point terylene harness and an Egyptian cotton Chamlon™ sweatband provide unrivalled comfort.
The safety cell concept prevents ricocheting debris travelling up the eyewear and impacting the scalp. In addition, the over spec cannot make contact with the scalp, preventing injury should it be forced back on to the safety cell. Optical class 1 over spec offers a panoramic view with minimal distortion. Conforms to EN166.1.FT offering impact protection at speeds of up to 100 mph (160 kph), tested using a 6mm steel ball bearing.
The optional clear plastic ID card holder fits standard business card size identification. Simple push and slide mechanism. A lamp bracket is positioned at the top of the unit.
Certificeringer: EN 397; EN 50365 Electrical Insulation.