Syringe pumps, VERITY®
Leverandør: GILSON
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
31130001 31130002 31130003
613-6999EA 36480 DKK
613-6999 613-7000 613-7001
Syringe pumps, VERITY®
Pumper Sprøjtepumper
The VERITY® syringe pumps provide speed and reliability to small- and large-volume, repetitive liquid transfers in HPLC, SPE, and other general liquid handling methods.
  • These pumps work with many VERITY® systems
  • Available in single syringe, dual syringe, and dual syringe with tee configurations
VERITY® 4000-series syringe pumps are ideal for all VERITY® GX-series liquid handlers and the 223 Sample Changer.
Bestillingsoplysninger: A pressure sensor is available in some models; contact your Avantor representative for more information.