Removable Deep Freeze Labels for Thermal Transfer Printers
Leverandør: GA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
RMTT-106C1-2OR RMTT-106C1-2WH RMTT-106C3-2OR RMTT-106C3-2WH RMTT-10C1-1WH RMTT-10C3-2WH RMTT-115C1-1WH RMTT-115C3-1WH RMTT-115C3-5WH RMTT-11C1-2WH RMTT-11C3-4WH RMTT122C10.3WH RMTT122C30.5WH RMTT-124C1-1WH RMTT-124C3-1WH RMTT-147C1-4WH RMTT-147C3-4WH RMTT-149C1-2WH RMTT-149C3-4WH RMTT-152C1-1WH RMTT-152C3-2WH RMTT-158C1-2WH RMTT-158C3-4WH RMTT-160C1-2WH RMTT-160C3-5WH RMTT164C1-10WH RMTT164C3-10WH RMTT-165C1-1WH RMTT-165C3-3WH RMTT-166C1-3WH RMTT166C3-10WH RMTT16710.25WH RMTT-19C1-1WH RMTT-19C3-1WH RMTT-203C1-1WH RMTT-203C3-1WH RMTT214NO110WH RMTT214NO310WH RMTT21910.25WH RMTT219C30.5WH RMTT231NPC11WH RMTT231NPC32WH RMTT-24C1-2WH RMTT-24C3-2WH RMTT-28C1-1OR RMTT-28C1-1WH RMTT-28C3-1OR RMTT-28C3-1WH RMTT-29C1-2WH RMTT-29C3-2WH RMTT-36C1-1WH RMTT-36C3-1WH RMTT-38C1-2WH RMTT-38C3-6WH RMTT-48C1-1WH RMTT-48C3-1WH RMTT-4C1-1WH RMTT-4C3-5WH RMTT52NOTC12WH RMTT52NOTC34WH RMTT533NP105WH RMTT533NPC31WH RMTT-58C1-1WH RMTT-58C3-2WH RMTT-5C1-1GA RMTT-5C1-1RE RMTT-5C1-1WH RMTT-5C1-1YE RMTT-5C3-3GA RMTT-5C3-3RE RMTT-5C3-3WH RMTT-5C3-3YE RMTT-70C1-2WH RMTT-70C3-4WH RMTT75NPC1-1WH RMTT75NPC3-1WH RMTT-7C1-1WH RMTT-7C3-3WH RMTT-94C1-1WH RMTT-94C3-1WH RMTT-97C1-1WH RMTT-97C3-1WH
GAINRMTT-106C1-2OREA 881 DKK
GAINRMTT-106C1-2OR GAINRMTT-106C1-2WH GAINRMTT-106C3-2OR GAINRMTT-106C3-2WH GAINRMTT-10C1-1WH GAINRMTT-10C3-2WH GAINRMTT-115C1-1WH GAINRMTT-115C3-1WH GAINRMTT-115C3-5WH GAINRMTT-11C1-2WH GAINRMTT-11C3-4WH GAINRMTT122C10.3WH GAINRMTT122C30.5WH GAINRMTT-124C1-1WH GAINRMTT-124C3-1WH GAINRMTT-147C1-4WH GAINRMTT-147C3-4WH GAINRMTT-149C1-2WH GAINRMTT-149C3-4WH GAINRMTT-152C1-1WH GAINRMTT-152C3-2WH GAINRMTT-158C1-2WH GAINRMTT-158C3-4WH GAINRMTT-160C1-2WH GAINRMTT-160C3-5WH GAINRMTT164C1-10WH GAINRMTT164C3-10WH GAINRMTT-165C1-1WH GAINRMTT-165C3-3WH GAINRMTT-166C1-3WH GAINRMTT166C3-10WH GAINRMTT16710.25WH GAINRMTT-19C1-1WH GAINRMTT-19C3-1WH GAINRMTT-203C1-1WH GAINRMTT-203C3-1WH GAINRMTT214NO110WH GAINRMTT214NO310WH GAINRMTT21910.25WH GAINRMTT219C30.5WH GAINRMTT231NPC11WH GAINRMTT231NPC32WH GAINRMTT-24C1-2WH GAINRMTT-24C3-2WH GAINRMTT-28C1-1OR GAINRMTT-28C1-1WH GAINRMTT-28C3-1OR GAINRMTT-28C3-1WH GAINRMTT-29C1-2WH GAINRMTT-29C3-2WH GAINRMTT-36C1-1WH GAINRMTT-36C3-1WH GAINRMTT-38C1-2WH GAINRMTT-38C3-6WH GAINRMTT-48C1-1WH GAINRMTT-48C3-1WH GAINRMTT-4C1-1WH GAINRMTT-4C3-5WH GAINRMTT52NOTC12WH GAINRMTT52NOTC34WH GAINRMTT533NP105WH GAINRMTT533NPC31WH GAINRMTT-58C1-1WH GAINRMTT-58C3-2WH GAINRMTT-5C1-1GA GAINRMTT-5C1-1RE GAINRMTT-5C1-1WH GAINRMTT-5C1-1YE GAINRMTT-5C3-3GA GAINRMTT-5C3-3RE GAINRMTT-5C3-3WH GAINRMTT-5C3-3YE GAINRMTT-70C1-2WH GAINRMTT-70C3-4WH GAINRMTT75NPC1-1WH GAINRMTT75NPC3-1WH GAINRMTT-7C1-1WH GAINRMTT-7C3-3WH GAINRMTT-94C1-1WH GAINRMTT-94C3-1WH GAINRMTT-97C1-1WH GAINRMTT-97C3-1WH
Removable Deep Freeze Labels for Thermal Transfer Printers
Etiketter
Removable deep-freeze labels for vials, tubes, and freezer boxes. These thermal transfer freezer labels have a glove-friendly adhesive that ensures the labels can be easily removed without leaving any adhesive residue.
  • Temperature range: –80 to +100 °C
  • Ideal for long-term storage in deep-freeze conditions (–80 °C)
  • Removable adhesive ensures the labels can be easily removed without leaving any adhesive residue
  • Printable with all major thermal-transfer printer brands
  • Resistant to gamma irradiation (tested up to 50 KGy)
Deep-freeze, removable barcode labels for vials, tubes, as well as polypropylene and cardboard boxes. The freezer labels have a strong, glove-friendly adhesive that allows them to adhere firmly to containers in (–20 , –40, –80 °C) freezers and dry ice while still being easily removable at room temperature without leaving behind any residue.
Waterproof and tearproof, these removable deep-freeze labels can also withstand temperatures ranging up to 100 °C, making them suitable for general lab use such as the identification of beakers, flasks, boxes, plastic and glass bottles that are stored in fridges and freezers.