Hagaki size permanent cryogenic labels for laser printers
Leverandør: GA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CLH-1A CLH-1BE CLH-1BL CLH-1BR CLH-1GA CLH-1GS CLH-1LA CLH-1LE CLH-1MT CLH-1OR CLH-1PI CLH-1RE CLH-1SI CLH-1VI CLH-1WH CLH-1YE CLH-2A CLH-2BE CLH-2BL CLH-2BR CLH-2GA CLH-2GS CLH-2LA CLH-2LE CLH-2MT CLH-2OR CLH-2PI CLH-2RE CLH-2SI CLH-2VI CLH-2WH CLH-2YE
GAINCLH-1AEA 303 DKK
GAINCLH-1A GAINCLH-1BE GAINCLH-1BL GAINCLH-1BR GAINCLH-1GA GAINCLH-1GS GAINCLH-1LA GAINCLH-1LE GAINCLH-1MT GAINCLH-1OR GAINCLH-1PI GAINCLH-1RE GAINCLH-1SI GAINCLH-1VI GAINCLH-1WH GAINCLH-1YE GAINCLH-2A GAINCLH-2BE GAINCLH-2BL GAINCLH-2BR GAINCLH-2GA GAINCLH-2GS GAINCLH-2LA GAINCLH-2LE GAINCLH-2MT GAINCLH-2OR GAINCLH-2PI GAINCLH-2RE GAINCLH-2SI GAINCLH-2VI GAINCLH-2WH GAINCLH-2YE
Hagaki size permanent cryogenic labels for laser printers
Etiketter
These labels can withstand deep-freezing and PCR thermocycler temperatures.
  • Temperature range: –196 to +120 °C
  • Multiple freeze-thaw cycles
Cryo-LazrTAG™ cryogenic plastic rectangle labels are designed for use with 0,2 ml PCR tubes, 0,5, 1,5, and 2 ml microcentrifuge tubes, and 1 to 2 ml cryo vials.