Signal cables, b.safe
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
R645-02 R643-12 R644-07 R644-12 R645-03 R645-05 R645-10
BOHLR645-02EA 220 DKK
BOHLR645-02 BOHLR643-12 BOHLR644-07 BOHLR644-12 BOHLR645-03 BOHLR645-05 BOHLR645-10
Signal cables, b.safe
Kabler
Cables connect waste caps with electronic fill level control or sensors to a b.safe display to monitor the fill level.