Waste pipes, b.safe
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
R1179-02 R1179-05 R1179-10
BOHLR1179-02EA 382 DKK
BOHLR1179-02 BOHLR1179-05 BOHLR1179-10
Waste pipes, b.safe
Fittings og stik
Straight tube made of static dissipative PE-EX.
Ext.Ø 32 mm suitable for b.safe Connecting tubes with clamp connections for waste pipes.