System waste caps LC, b.safe
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
R1305-01 R1306-01 R1309-01
BOHLR1305-01EA 3060 DKK
BOHLR1305-01 BOHLR1306-01 BOHLR1309-01
System waste caps LC, b.safe
Låg Sikkerhedslåg og sikkerhedsaffaldslåg
b.safe waste tubes can be connected to the canister by means of these b.safe system waste caps and solvents can be collected safely. To avoid spill over of canisters through the connection of a Waste Cap to a b.safe Display for electronic level control. The filling levels of waste containers in laboratory cupboards and workbenches can be reliably monitored. With the use of b.safe Waste Tubes, solvent waste is led directly into the canister, solvent vapours are held back by the exhaust filter with activated carbon or can be led to the hood by means of a venting hose.
  • Made of high-performance plastics
  • Thermally and chemically stable
  • Good grip for easy assembly, safe working and improved processes
Screw cap made of PPS or PP (only canister thread S90), free movable insert made of conductible PE-EX with permanently fixed cable (length approx. 0,5 m) for the connection to a b.safe Display as well as sight glass and visual pin for additional optic control of the filling level. With 1 hose connector GL 25 suitable for b.safe waste tubes and 1 connector GL 14 for fixing the b.safe Venting Hose or a b.safe exhaust filter.
Design and scope of delivery according to table.