Modular vacuum concentrators, Savant™ SpeedVac™
Leverandør: THERMO SAVANT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SPD210-230 SPD210P1-230 SPD210P2-230
THSASPD210-230EA 70280 DKK
THSASPD210-230 THSASPD210P1-230 THSASPD210P2-230
Modular vacuum concentrators, Savant™ SpeedVac™
Fordampere Centrifugalfordampere
The Thermo Scientific™ Savant™ SpeedVac™ SPD210 vacuum concentrator is a large-capacity vacuum concentrator with a modular design, ideal for drying aqueous and organic solvents.
  • Kit
  • Medium capacity
  • Variety of rotors
  • Glass safety cover
  • PTFE coated aluminum chamber
The SpeedVac™ SPD210 is capable of drying larger quantities of samples at one time and will support your high-volume sample preparation needs.
Bestillingsoplysninger: Supplied as different packages, please see description for details of what is included in specific kits/packages.