ESD safety shoes, lace-up, jo_Slim Low, Jori
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
12831/36 12831/37 12831/38 12831/39 12831/40 12831/41 12831/42 12831/43 12831/44 12831/45 12831/46 12831/47 12831/48 12841/36 12841/37 12841/38 12841/39 12841/40 12841/41 12841/42 12841/43 12841/44 12841/45 12841/46 12841/47 12841/48 12851/36 12851/37 12851/38 12851/39 12851/40 12851/41 12851/42 12851/43 12851/44 12851/45 12851/46 12851/47 12851/48
ELTN12831/36EA 462 DKK
ELTN12831/36 ELTN12831/37 ELTN12831/38 ELTN12831/39 ELTN12831/40 ELTN12831/41 ELTN12831/42 ELTN12831/43 ELTN12831/44 ELTN12831/45 ELTN12831/46 ELTN12831/47 ELTN12831/48 ELTN12841/36 ELTN12841/37 ELTN12841/38 ELTN12841/39 ELTN12841/40 ELTN12841/41 ELTN12841/42 ELTN12841/43 ELTN12841/44 ELTN12841/45 ELTN12841/46 ELTN12841/47 ELTN12841/48 ELTN12851/36 ELTN12851/37 ELTN12851/38 ELTN12851/39 ELTN12851/40 ELTN12851/41 ELTN12851/42 ELTN12851/43 ELTN12851/44 ELTN12851/45 ELTN12851/46 ELTN12851/47 ELTN12851/48
ESD safety shoes, lace-up, jo_Slim Low, Jori
Sko og støvler
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Padded tongue
  • Rubber/EVA/EVA sole jo_SAMBA
  • Steel toe cap
Suede/hydrophobised textile material construction with fabric lining for natural ventilation. Full-length inlay ESD sole. Non-metallic, penetration-resistant midsole.
Certificeringer: EN ISO 20345 S3 SRC, Form A.