ESD safety shoes, lace-up, S1P, Marten XXSports Low
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
728121/35 728121/36 728121/37 728121/38 728121/39 728121/40 728121/41 728121/42 728121/43 728121/44 728121/45 728121/46 728121/47 728121/48
ELTN728121/35EA 701 DKK
ELTN728121/35 ELTN728121/36 ELTN728121/37 ELTN728121/38 ELTN728121/39 ELTN728121/40 ELTN728121/41 ELTN728121/42 ELTN728121/43 ELTN728121/44 ELTN728121/45 ELTN728121/46 ELTN728121/47 ELTN728121/48
ESD safety shoes, lace-up, S1P, Marten XXSports Low
Sko og støvler
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Full, padded bellows tongue
  • TPU/PU sole WELLMAXX SPORTS
  • Steel toe cap
  • Extra laces included
Microfibre/mesh material construction with fabric lining for natural ventilation. Full-length inlay sole, SPORTIVE ESD. Non-metallic, penetration-resistant midsole. Lacing system with individual quick locking system. Sole core made of Infinergy® by BASF.
Certificeringer: EN ISO 20345 S1P SRA, form A.