ESD safety shoes, lace-up, Larkin XXSports Low
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
728110/35 728110/36 728110/37 728110/38 728110/39 728110/40 728110/41 728110/42 728110/43 728110/44 728110/45 728110/46 728110/47 728110/48
ELTN728110/35EA 656 DKK
ELTN728110/35 ELTN728110/36 ELTN728110/37 ELTN728110/38 ELTN728110/39 ELTN728110/40 ELTN728110/41 ELTN728110/42 ELTN728110/43 ELTN728110/44 ELTN728110/45 ELTN728110/46 ELTN728110/47 ELTN728110/48
ESD safety shoes, lace-up, Larkin XXSports Low
Sko og støvler
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Padded tongue
  • TPU/PU sole WELLMAXX SPORTS
  • Steel toe cap
  • Leather-free
Mesh material construction with fabric lining for natural ventilation. Full-length inlay sole, SPORTIVE ESD. Antistatic soft fleece insole. Sole core made of Infinergy® by BASF.
Certificeringer: EN ISO 20345 S1 SRA, form A.