ESD easy safety sandals, Velcro® fastening, S1P, Impulse Lady XXTL Low
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
741301/35 741301/36 741301/37 741301/38 741301/39 741301/40 741301/41 741301/42
ELTN741301/35EA 664 DKK
ELTN741301/35 ELTN741301/36 ELTN741301/37 ELTN741301/38 ELTN741301/39 ELTN741301/40 ELTN741301/41 ELTN741301/42
ESD easy safety sandals, Velcro® fastening, S1P, Impulse Lady XXTL Low
Sko og støvler
Ladies' safety sandal is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Padded tongue
  • PU/PU sole WELLMAXX TRAINERS LADY
  • Steel toe cap
  • Leather-free
  • Velcro® closure
Microfibre construction with fabric lining for natural ventilation. Full-length inlay ESD sole, PRO Lady Y. Non-metallic, penetration-resistant midsole. Sole core made of Infinergy® by BASF.
Certificeringer: EN ISO 20345 S1P SRC, form A.
Bestil nu
Specifikationer
Environmentally Preferable Lav fremstillingspåvirkning
Miljøcertificering ISO 14001