ESD safety shoes, Boa® Fit System, S3, jo_Swift BOA® Low
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
12081/36 12081/37 12081/38 12081/39 12081/40 12081/41 12081/42 12081/43 12081/44 12081/45 12081/46 12081/47 12081/48
ELTN12081/36EA 649 DKK
ELTN12081/36 ELTN12081/37 ELTN12081/38 ELTN12081/39 ELTN12081/40 ELTN12081/41 ELTN12081/42 ELTN12081/43 ELTN12081/44 ELTN12081/45 ELTN12081/46 ELTN12081/47 ELTN12081/48
ESD safety shoes, Boa® Fit System, S3, jo_Swift BOA® Low
Sko og støvler
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Padded tongue
  • Rubber/PU sole jo_ATHLETIC
  • Composite toe cap
  • BOA® Fit System
  • Leather-free
Hydrophobised microfibre construction with fabric lining for natural ventilation. Full-length inlay ESD sole. Non-metallic, penetration-resistant midsole.
Certificeringer: EN ISO 20345 S3 HRO/SRC, form A.