ESD safety ankle boots, lace-up, Marten XXSports Mid
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
768121/35 768121/36 768121/37 768121/38 768121/39 768121/40 768121/41 768121/42 768121/43 768121/44 768121/45 768121/46 768121/47 768121/48
ELTN768121/35EA 716 DKK
ELTN768121/35 ELTN768121/36 ELTN768121/37 ELTN768121/38 ELTN768121/39 ELTN768121/40 ELTN768121/41 ELTN768121/42 ELTN768121/43 ELTN768121/44 ELTN768121/45 ELTN768121/46 ELTN768121/47 ELTN768121/48
ESD safety ankle boots, lace-up, Marten XXSports Mid
Sko og støvler
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Full, padded bellows tongue
  • TPU/PU sole, WELLMAXX SPORTS
  • Steel toe cap
  • Leather-free
  • Extra laces included
Microfibre/textile material construction with fabric lining for natural ventilation. Full-length inlay sole, SPORTIVE ESD. Non-metallic, penetration-resistant midsole. Sole core made of Infinergy® by BASF.
Colour: Blue
Certificeringer: EN ISO 20345 S1P SRA, Form B.