ESD safety shoes, lace-up, jo_Free Low, Jori
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
12011/36 12011/37 12011/38 12011/39 12011/40 12011/41 12011/42 12011/43 12011/44 12011/45 12011/46 12011/47 12011/48
ELTN12011/36EA 500 DKK
ELTN12011/36 ELTN12011/37 ELTN12011/38 ELTN12011/39 ELTN12011/40 ELTN12011/41 ELTN12011/42 ELTN12011/43 ELTN12011/44 ELTN12011/45 ELTN12011/46 ELTN12011/47 ELTN12011/48
ESD safety shoes, lace-up, jo_Free Low, Jori
Sko og støvler
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Padded tongue
  • Rubber/PU sole jo_ATHLETIC
  • Composite toe cap
  • Metal- and leather-free
Hydrophobised microfibre construction with fabric lining for natural ventilation. Full-length inlay ESD sole. Non-metallic, penetration-resistant midsole.
Certificeringer: EN ISO 20345 S3 HRO/SRC, Form A.