ESD safety shoes, lace-up, Marty XXSports Low
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
728181/40 728181/41 728181/42 728181/43 728181/44 728181/45 728181/46 728181/47 728181/48
ELTN728181/40EA 679 DKK
ELTN728181/40 ELTN728181/41 ELTN728181/42 ELTN728181/43 ELTN728181/44 ELTN728181/45 ELTN728181/46 ELTN728181/47 ELTN728181/48
ESD safety shoes, lace-up, Marty XXSports Low
Sko og støvler
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Padded tongue
  • TPU/PU sole WELLMAXX SPORTS
  • Steel toe cap
  • Leather-free
Hydrophobised CORDURA® textile material construction with fabric lining for natural ventilation. Full-length inlay SPORTIVE ESD sole. Non-metallic, penetration-resistant midsole. Sole core made of Infinergy® by BASF.
Colour: Black/white/blue
Certificeringer: EN ISO 20345 S3 SRA, Form A.