ESD safety shoes, lace-Up, S3, jo_Rapid Low, Jori
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
12021/36 12021/37 12021/38 12021/39 12021/40 12021/41 12021/42 12021/43 12021/44 12021/45 12021/46 12021/47 12021/48 12031/36 12031/37 12031/38 12031/39 12031/40 12031/41 12031/42 12031/43 12031/44 12031/45 12031/46 12031/47 12031/48
ELTN12021/36EA 485 DKK
ELTN12021/36 ELTN12021/37 ELTN12021/38 ELTN12021/39 ELTN12021/40 ELTN12021/41 ELTN12021/42 ELTN12021/43 ELTN12021/44 ELTN12021/45 ELTN12021/46 ELTN12021/47 ELTN12021/48 ELTN12031/36 ELTN12031/37 ELTN12031/38 ELTN12031/39 ELTN12031/40 ELTN12031/41 ELTN12031/42 ELTN12031/43 ELTN12031/44 ELTN12031/45 ELTN12031/46 ELTN12031/47 ELTN12031/48
ESD safety shoes, lace-Up, S3, jo_Rapid Low, Jori
Sko og støvler
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Padded tongue
  • Rubber/PU sole jo_ATHLETIC
  • Rubber scuff cap
  • Composite toe cap
  • Metal- and leather-free
  • Available in multiple colour options
Hydrophobised microfibre construction with fabric lining for natural ventilation. Full-length inlay ESD sole. Non-metallic, penetration-resistant midsole.
Certificeringer: EN ISO 20345 S3 HRO/SRC, form A.