ESD safety shoes, Boa® Fit System, S1P, jo_Speedy BOA® Low, Jori
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
12171/36 12171/37 12171/38 12171/39 12171/40 12171/41 12171/42 12171/43 12171/44 12171/45 12171/46 12171/47 12171/48
ELTN12171/36EA 589 DKK
ELTN12171/36 ELTN12171/37 ELTN12171/38 ELTN12171/39 ELTN12171/40 ELTN12171/41 ELTN12171/42 ELTN12171/43 ELTN12171/44 ELTN12171/45 ELTN12171/46 ELTN12171/47 ELTN12171/48
ESD safety shoes, Boa® Fit System, S1P, jo_Speedy BOA® Low, Jori
Sko og støvler
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Padded tongue
  • Rubber/EVA sole jo_DYNAMIC
  • Composite toe cap
  • BOA® Fit System
  • Leather-free
Mesh material/microfibre fabric construction with lining for natural ventilation. Full-length inlay ESD sole. Non-metallic, penetration-resistant midsole.
Certificeringer: EN ISO 20345 S1P HRO/SRC, form A.