ESD safety shoes, BOA® Fit System, HANNAH XXTL BOA® Low
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
742561/35 742561/36 742561/37 742561/38 742561/39 742561/40 742561/41 742561/42
ELTN742561/35EA 753 DKK
ELTN742561/35 ELTN742561/36 ELTN742561/37 ELTN742561/38 ELTN742561/39 ELTN742561/40 ELTN742561/41 ELTN742561/42
ESD safety shoes, BOA® Fit System, HANNAH XXTL BOA® Low
Sko og støvler
Ladies' safety shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Full, padded bellows tongue
  • PU/PU sole, WELLMAXX TRAINERS LADY
  • Steel toe cap
  • BOA® Fit System
  • Leather-free
Hydrophobised microfibre/hydrophobised textile material construction with fabric lining for natural ventilation.Full-length inlay sole, ESD PRO Lady Y.Non-metallic, penetration-resistant midsole.Sole core made of Infinergy® by BASF.
Certificeringer: EN ISO 20345 S3 SRC, form A.