Floor cleaners, SURE®
Leverandør: Diversey
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
100891586 100891588
141-1571EA 505 DKK
141-1571 141-1572
Floor cleaners, SURE®
Renseopløsninger Detergenter og Overflade Rengøringsmidler
Concentrated, low foaming daily cleaner for all water resistant hard floors. Recommended for all cleaning methods, including bucket and scrubber drier applications.
  • Concentrated formulation gives excellent economy of use
  • No residues on the floor and no build-up
  • Low foaming effective cleaning
  • No fragrance or dye
  • Contains plant-based ingredients from renewable sources
  • Decomposed by natural processes
  • No environmental hazard classifications, no safety warnings for user
SURE® floor cleaner is part of a unique range of plant-based, cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance.
Certificeringer: OECD 301B.
EU-Ecolabel (EU: NL/020/045).
The surfactants used in this product are in compliance with the requirement of EU Detergent Regulation, EC 648/2004.