Hand dishwash, SURE®
Leverandør: Diversey
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
101100604 101100583
115-3100EA 452 DKK
115-3100 115-3099
Hand dishwash, SURE®
Renseopløsninger Detergenter og Overflade Rengøringsmidler
Concentrated hand dishwashing liquid for cleaning pots, pans, crockery, glass and all washable utensils.
  • No fragrance or dye (EU Ecolabel requirement)
  • Lesser foaming (suitable for pre-soak before machine ware washing)
  • Contains plant-based ingredients from renewable sources
  • Ddecomposed by natural processes
  • No environmental hazard classifications, no safety warnings for user
  • The product is non classified
  • Concentrated formulation gives excellent economy in use
  • Streak-free, easy rinsing for a shiny finish
  • Bottles/cans suitable for recycling
SURE® hand dishwash is part of a unique range of plant-based, cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance.
Certificeringer: OECD 301B test
EU Ecolabel (EU: NL/019/012)
The surfactants used in this product are in compliance with the requirement of EU Detergent Regulation, EC 648/2004