Descalers, SURE®
Leverandør: Diversey
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
100892014 100892015
141-1584EA 460 DKK
141-1584 141-1585
Descalers, SURE®
Renseopløsninger Detergenter og Overflade Rengøringsmidler
Concentrated acidic cleaner for removal of lime scale from kitchen equipment and utensils.
  • Specially developed for frequent descaling
  • Excellent adherence on vertical surfaces
  • Very efficient on micro incrustations and mineral soiling
  • No fragrance or dye
  • Contains plant-based ingredients from renewable sources
  • Decomposed by natural processes
  • No environmental hazard classifications, no safety warnings for product in use for user
SURE® descaler is part of a unique range of plant-based, cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance.
Certificeringer: OECD 301B test, Cradle to cradle certification at gold level.