Filling needle autoclave bags with drawstring, Tyvek® 1422A
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BGT2A0101D BGT2A0102D BGT2A0103D BGT2A0103D10 BGT2A0104D BGT2A0105D BGT2A0106D BGT2A0107D BGT2A0108D BGT2A0111D BGT2A0202D BGT2A0202D16 BGT2A0203D BGT2A0204D BGT2A0205D BGT2A0206D BGT2A0207D BGT2A0208D BGT2A0209D BGT2A0210D BGT2A0211D BGT2A0212D BGT2A0213D BGT2A02503D BGT2A0303D18 BGT2A0304D BGT2A0305D BGT2A0306D BGT2A0308D BGT2A0309D BGT2A0310D BGT2A0312D BGT2A0314D BGT2A0403D BGT2A0404D BGT2A0410D
129-3374EA 21490 DKK
129-3374 129-3375 129-3376 129-3377 129-3378 129-3380 129-3382 129-3383 129-3385 129-3387 129-3389 129-3188 129-3190 129-3191 129-3193 129-3195 129-3197 129-3199 129-3200 129-3201 129-3202 129-3203 129-3204 129-3207 129-3208 129-3209 129-3210 129-3214 129-3215 129-3217 129-3219 129-3220 129-3221 129-3222 129-3170 129-3165
Filling needle autoclave bags with drawstring, Tyvek® 1422A
Poser Steriliseringsposer
Protect your filling needles from accidental touching after they are removed from the primary sterilisation packaging. Also creates secondary level of protection during aseptic presentation.
  • Tear and puncture resistant
  • Eliminates need to cut material and the use of autoclave tape
  • Fast and consistent for equipment preparation area
  • Steam indicator
  • Hydrophobic and lint-free
  • Reliable factory seals
  • Exceptional microbial barrier
These Tyvek® filling needle bags stay in place via the adjustable drawstring closure system. The bags can be aseptically removed with forceps after the filling needles are set in place, without having to cut, or remove particulate generating autoclave tape. These are manufactured and packaged in a controlled cleanroom environment.
Leveringsinformation: Certificates of Conformance sent with each shipment.