Stopper Bags with Steam Indicator, Tyvek® 1073B/HDPE
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SBMGTVKHS0304 SBMGTVKHS0306 SBMGTVKHS0409 SBMGTVKHS0422 SBMGTVKHS0508 SBMGTVKHS0610 SBMGTVKHS0630 SBMGTVKHS0812 SBMGTVKHS1035 SBMGTVKHS1212 SBMGTVKHS1215 SBMGTVKHS1219 SBMGTVKHS1238 SBMGTVKHS1318 SBMGTVKHS1414 SBMGTVKHS1518 SBMGTVKHS1616 SBMGTVKHS1632 SBMGTVKHS1824 SBMGTVKHS1920 SBMGTVKHS2022 SBMGTVKHS2026 SBMGTVKHS2216 SBMGTVKHS2418 SBMGTVKHS2436 SBMGTVKHS2440 SBMGTVKHS2848 SBMGTVKSS1214 1632SBSD
129-2910EA 10080 DKK
129-2910 129-2911 129-2921 129-2922 129-2923 129-2924 129-2925 129-2926 129-2927 129-2928 129-2929 129-2930 129-2931 129-2932 129-2933 129-2934 129-2935 129-2936 129-2937 129-2938 129-3014 129-3015 129-3016 129-3017 129-3018 129-3019 129-3022 129-3023 KACC1632SBSD
Stopper Bags with Steam Indicator, Tyvek® 1073B/HDPE
Poser Steriliseringsposer
Tyvek® 1073B/HDPE autoclave bags are ideal for steam sterilisation of pharmaceutical components such as vial and syringe stoppers and caps. Also a good choice for heavy equipment parts where visibility into the bag is required.
  • Manufactured and packaged in a controlled cleanroom environment
  • Steam sterilisation indicator - autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • One side made of hydrophobic and lint-free Tyvek® 1073B
  • One side made of semi-transparent HDPE film
  • Excellent impact resistance with exceptional microbial barrier
  • Consistently strong, double welded factory seals
Tyvek® 1073B side permits steam penetration during sterilisation. These are a good choice for heavy, or pharmaceutical change parts with sharp corners or where visibility into the bag is required after sterilisation.
Certificeringer: Supplied with Certificates of Conformance.