Autoclave Bags with Steam Indicator and Tyvek® Drawstring Closure, BHD
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BNWBG0606D2P BNWBG0642D2P BNWBG09.512D2P BNWBG1212D2P BNWBG1218D2P BNWBG1624D2P BNWBG01644D2P BNWBG1830D2P BNWBG1854D2P BNWBG2024D2P BNWBG2032D2P BNWBG2424D2P BNWBG2435D2P BNWBG2436D2P BNWBG3648D2P BNWBG4040D2P BNWT3654D22P
129-2776EA 53270 DKK
129-2776 129-2777 129-2778 129-2779 129-2780 129-2781 129-2770 129-2782 129-2783 129-2784 129-2785 129-2786 129-2787 129-2788 129-2789 129-2790 129-2791
Autoclave Bags with Steam Indicator and Tyvek® Drawstring Closure, BHD
Poser Steriliseringsposer
BHD autoclave bags with Tyvek® drawstrings can be used as either the inner, or outer bag for the steam sterilisation of pharmaceutical change parts.
  • Manufactured and packed in a controlled cleanroom environment
  • Reduces labour costs
  • Simplifies conformance when wrapping to SOP's
  • Steam sterilisation indicator - autoclavable up to 135 °C
  • Tear and puncture resistant 2 ply blue BHD non woven
  • Significant process improvement compared to wrapping with sheets and tape
Standardise and simplify the way equipment is wrapped for steam sterilisation. The use of these 2 ply drawstring bags will also save valuable time in the equipment prep area. The 2 ply BHD material resists ripping and punctures.
Since 100% of the surface area allows for steam penetration and drying, each load can be configured for maximum efficiency and throughput. The drawstring bags can be aseptically opened without generating particles like cutting other types of bags open can.
Certificeringer: Certificates of Conformance sent with each shipment.