Filling Needle Autoclave Bags with Steam Indicator and Tyvek® Drawstring Closure, BHD
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BNWBG0104D2P BNWBG0106D2P BNWBG0108D2P BNWBG0204D2P BNWBG0206D2P BNWBG0210D2P BNWBG0308D2P BNWBG0310D2P
129-2767EA 46930 DKK
129-2767 129-2768 129-2769 129-2771 129-2772 129-2773 129-2774 129-2775
Filling Needle Autoclave Bags with Steam Indicator and Tyvek® Drawstring Closure, BHD
Poser Steriliseringsposer
These bags protects needles from accidental touching after they are removed from the primary sterilisation packaging.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labour costs
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilisation indicator - autoclavable up to 135 °C
  • Tear and puncture resistant 2-ply blue BHD non-woven
  • Significant process improvement compared to wrapping with sheets and tape
These BHD filling needle bags stay in place via the adjustable Tyvek® drawstring closure system. The bags can be aseptically removed with forceps after the filling needles are set in place, without having to cut, or remove particulate generating autoclave tape.
Certificeringer: Certificates of Conformance sent with each shipment.