Cleanroom gloves, nitrile, sterile, PUREZERO® HG3
Leverandør: Halyard
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CLN923210 CLN923260 CLN923265 CLN923270 CLN923275 CLN923280 CLN923285 CLN923290
112-6494EA 4200 DKK
112-6494 112-6495 112-6496 112-6497 112-6498 112-6499 112-6500 112-6501
Cleanroom gloves, nitrile, sterile, PUREZERO® HG3
Handsker
High quality gloves that are designed to exact standards to meet the requirements of a contamination controlled environment.
  • High quality nitrile
  • Sterile
  • Accelerator-free formula reduces risk of type IV allergies
  • For use in cleanroom environments (ISO 4/5, grade A/B aseptic)
  • REACH Annex XVII
Offering the right glove for the right controlled environment, PUREZERO® provides a cleanroom glove with guaranteed low particulate counts and endotoxin levels due to repeated washes in deionised water and chlorination. Nickel-, powder-, silicon- and latex-free, with no phthalates. These gloves are formulated with zero presence of these hazardous traditional accelerators: Sulphur, thiurams, thiazoles, guanidine and carbamates (ZDEC or ZDBC). All backed up by a vertically integrated supply chain.
Certificeringer: These cleanroom gloves are certified as PPE category III as per Regulation (EU) 2016/425. The applicable standards are:
• EN ISO 374-1:2016 Type C K - dangerous chemicals and microorganisms (permeation time)
• EN ISO 374-2:2019 - dangerous chemicals and microorganisms (determining resistance to penetration holes/defects)
• EN ISO 374-4:2019 - resistance to degradation by chemicals
• EN ISO 374-5:2016 - microorganism and virus protection
• EN 420:2003 + A1:2009 - general requirements for protective gloves
• EN 16523-1:2015 + A1:2018 - permeation of chemicals (+IPA 70%**)
• ISO 16604:2004 - contact with blood and body fluids
• ASTM D6978 - permeation testing on chemotherapy drugs (14 drugs)
Emballering: Gloves are pair packed in poly wallet pouches.