Anti-static explosion-proof tubing rolls, PTFE EX, BOLA
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S2040-32 S2040-40 S2040-50 S2040-60 S2040-64
BOHLS2040-32EA 675 DKK
BOHLS2040-32 BOHLS2040-40 BOHLS2040-50 BOHLS2040-60 BOHLS2040-64
Anti-static explosion-proof tubing rolls, PTFE EX, BOLA
Slanger Overførselsslange
Very good electric conductivity due to a special 'anti-static compound' made of pure PTFE and highly pure carbon dust (less than 2,5%).
  • Extensive chemical resistance due to PTFE parts
  • Resistance of less than 1000 kOhm according to DIN EN 12115 directive
  • Ideal for light-sensitive substances
Use for anti-static applications where explosion protection is required, particularly for the transport of solvents or alcohols.
Colour: black