Autoclavable Muff Covers, DuPont™ Tyvek®
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFT2A03 MFT2A04 MFT2A05 MFT2A05OT MFT2A06BT MFT2A07 MFT2A08 MFT2A09 MFT2A12 MFT2A16 MFT2A18 1034TW
129-3076EA 19180 DKK
129-3076 129-3077 129-3078 129-3123 129-3125 129-3126 129-3128 129-3129- 129-3131 129-3133 129-3134 129-4200
Autoclavable Muff Covers, DuPont™ Tyvek®
Covers
Save time and standardise the activities in equipment preparation for sterilisation.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labour costs
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilisation indicator - autoclavable at 121 to 125 °C
  • Tear- and puncture-resistant, hydrophobic and lint-free Tyvek® 1422A
  • With elastic to fit a range of diameters
  • Conveniently packaged 100 per sealed poly bag
Tyvek® Muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes, and hoses. These closures are breathable and can be used to prevent particulates and microbial contamination from entering openings of any size. Since they are autoclavable, they can be used during sterilisation to reduce time consuming wrapping of open containers. Each closure incorporates latex-free rubber elastic to create a positive seal around round shaped openings.
Certificeringer: Certificates of conformance sent with each shipment.