Autoclavable muff covers with skirt, Tyvek®
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFT2A05SK02 MFT2A07SK02 MFT2A09SK02 MFT2A12SK02 MFT2A18SK02 MFT2A18SK02TL
129-3124EA 50580 DKK
129-3124 129-3127 129-3130 129-3132 129-3135 129-3136
Autoclavable muff covers with skirt, Tyvek®
Covers
Save time and standardize the activities in equipment preparation for sterilisation.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labour costs
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilization indicator - autoclavable at 121 to 125 °C
  • Tear- and puncture-resistant, hydrophobic and lint-free Tyvek® 1422A
  • With elastic to fit a range of diameters
  • With skirt extension for easy removal
  • Conveniently packaged 100 per sealed poly bag
Save time and standardize the activities in equipment preparation for sterilization. Tyvek® Muffs with a skirt extension provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes, and hoses. These closures are breathable and can be used to prevent particulates and microbial contamination from entering openings of any size. Since they are autoclavable, they can be used during sterilization to reduce time consuming wrapping of open containers. Each closure incorporates latex-free rubber elastic to create a positive seal around round shaped openings with the addition of a 2" (51 mm) long skirt extension. The skirt extension provides a safe area to remove the muff without inadvertently touching the critical surfaces with the fingers.
Certificeringer: Certificates of conformance sent with each shipment.