Autoclavable Stopper Bowl Covers, DuPont™ Tyvek®
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BWLT2A20 BWLT2A24 BWLT2A26 BWLT2A30 BWLT2A34 BWLT2A36 BWLT2A38 BWLT2A42 BWLT2A46 BWLT2A48 BWLT2A54 BWLT2A56 BWLT2A60 BWLT2A64 BWLT2A72 BWLT2A90 BWLT2A120 BWLT2A144 BWLT2A52 BWLT2A32 BWLT2A36-2PLY8
129-2880EA 34400 DKK
129-2880 129-2881 129-2882 129-2883 129-2885 129-2886 129-2887 129-2888 129-2889 129-2890 129-2892 129-2893 129-2894 129-2895 129-2896 129-2897 129-2878 129-2879 129-2891 129-2884 129-3989
Autoclavable Stopper Bowl Covers, DuPont™ Tyvek®
Covers
Save time and standardise activities in equipment preparation for sterilisation. Leave in place during stopper bowl installation to prevent microbial and particulate contamination, and while remaining fill line set-up activities are performed.
  • Manufactured and packed in a controlled cleanroom environment
  • Reduces labour costs
  • Individually poly bagged
  • Simplifies conformance when wrapping to SOP's
  • Steam sterilisation indicator - autoclavable at 121 to 125 °C
  • Elastic to fit a range of diameters
  • Tear and puncture resistant, hydrophobic, and lint-free Tyvek® 1422A
Stopper bowl covers provide a convenient, time-saving method of wrapping stopper bowls prior to steam sterilisation. These covers allow immediate steam penetration and work best with modern pulse vacuum autoclaves for sufficient drying.
After sterilisation, and after the outer Tyvek® bag is removed, leave cover in place during installation to prevent microbial and particulate contamination. Remove after all other filling line set-up activities are completed. Remaining in place over the bowl during set-up, their use minimises microbial or particulate contamination risks.
Certificeringer: Certificates of Conformance sent with each shipment.