VWR® Petri Dishes, Glass
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
391-0652EA 283 DKK
391-0652 391-0660 391-0665 391-0668 391-0671 391-0673 391-0674 391-0698
VWR® Petri Dishes, Glass
Dyrkningsplader Petriskåle
Soda-lime glass, with lid. For use as disposables. These Petri dishes are a green alternative to disposable Petri dishes made from plastic.
  • Good chemical resistance
  • Heatable up to 60 °C
  • Not suitable for applications related to strong heat or rapid temperature changes
  • Ideal for bacterial culture production
Advarsel: The indication of the diameter of the petri dishes is approximate.