Reaktor med omrøring, GL 45, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1200395 1200396 1200390 1200391
215-2359EA 3310 DKK
215-2359 215-2360 215-2357 215-2358
Reaktor med omrøring, GL 45, DURAN®
Kolber Spinnerflasker
DURAN®, borosilicate glass 3.3, clear, PP, PTFE, PEEK, stainless steel.
  • Significantly improved mixing compared to standard magnetic stirring bar
  • Drive using standard magnetic stirrers
  • Variable stirrer shaft, can be used for 500 and 1000 ml DURAN® GL 45 laboratory bottles
  • Can be used up to 500 rpm
  • Parts in contact with media conform fully to FDA requirements
  • Temperature resistant up to 140 °C
The DURAN® stirred reactor is ideal for the widest possible range of mixing processes in laboratories, e.g. mixing liquids or dissolving solids. The connectionsMore Product Information