Calibrated thermocouple thermometers, H-B Durac®
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B609010800 B609011000 B609011100 B609110200
620-2817EA 3210 DKK
620-2817 620-2818 620-2819 620-2820
Calibrated thermocouple thermometers, H-B Durac®
Termometre Digital termometre
Used for temperature measurement in life science research, kilns, gas turbine exhaust, diesel engines, and other industrial processes, H-B calibrated thermocouple thermometers provide measurement throughout a broad temperature range and the ability to utilise multiple probes at the same time.
  • Wide temperature range allows accurate monitoring at extreme temperatures
  • Max./min./average temperature recording capability
  • Data hold
  • Software CD (where included) for real time data collection and logging via USB
Calibration points: 0 °C, 100 °C.
Certificeringer: Tested and calibrated in H-B’s exclusive triple-accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST. Individually serialised, CE-marked. Note: Calibration applies to included probe and tested port only.
Bestillingsoplysninger: Probes for wider temperature ranges available (VWRI620-2821 and VWRI620-2822) or use alternatively purchased probes; see chart for probe types accepted by each unit.
Leveringsinformation: Includes two K-type beaded wire probes (–20 to +250 °C capability); one is calibrated and serialised.