NHS Mag Sepharose
Leverandør: Cytiva
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
28-9440-09 28-9513-80
28-9440-09EA 365.52 DKK
28-9440-09 28-9513-80
NHS Mag Sepharose
Kugler Magnetiske perler
NHS Mag Sepharose magnetic beads are designed for pull-down techniques, involving isolation of a protein through adsorption of complex samples onto beads, enabling rapid capture and enrichment of selected proteins based on affinity.
  • Visible and dense Sepharose based magnetic beads with N-hydroxysuccinimide ligand for easy coupling of biomolecules containing a free amine group
  • Non-adherent beads eliminate smearing effects and aggregate formation
  • Can be used without detergents
  • Simple capture of target protein in small or large sample volumes (low microliter