Electric Bunsen Burners, LAB 2, LAB 3
Leverandør: K-Factory
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
LAB3 LAB2
766-0102EA 5360 DKK
766-0102 766-0101
Electric Bunsen Burners, LAB 2, LAB 3
Varmeapparater Elektrisk brænder
Heating process based on a patented hot air column system. The burner has the same applications as bunsen burners.
  • Maximum temperatures of 650 °C (LAB2) and 750 °C (LAB3)
  • The sides stay cool, even after many operating hours
  • Integrated power regulator
  • Excellent isolated housing
  • Protection against accidental liquid spillage
Power: 500 W (LAB2), 900 W (LAB3)
W×D×H: 149×168×192 mm