Micro-dispensers, Piccolo 1 and 2
Leverandør: VITLAB
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1610501 1611506 1610503 1610504 1610502 1610506 1611503 1611508
613-1254EA 2380 DKK
613-1254 613-1260 613-1256 613-1257 613-1255 613-1258 613-1259 613-1261
Micro-dispensers, Piccolo 1 and 2
Flasketopdispenser
For dispensing tiny quantities of liquids in all areas of biochemical and medical research.
  • Reset mechanism refills the volume automatically
  • Disposable tips are unnecessary
  • Can be operated with only one hand
  • Use the thumb to depress the volume dispensing button, just as with a pipette
  • Discharge tube can be rotated over 360° so that it is always optimally situated with respect to the bottle label
The Piccolo are designed for serial dispensing operations. In order to guarantee high resistance to chemicals and a broad range of applications, only high-quality materials, such as PTFE, PFA, ETFE, FEP, borosilicate glass, and platinum-iridium come in contact with the media.
Leveringsinformation: Supplied with mounting tools and an instruction manual.