Glogos II autorad markers
Leverandør: AGILENT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
420201 420202 420203
AGLS420201EA 1350 DKK
AGLS420201 AGLS420202 AGLS420203
Glogos II autorad markers
Proteinkoblings- og mærkningssystemer
Luminescent peel-off stickers are used for accurately marking and aligning autoradiographs.
  • Adaptable for long or short exposures
Easily affixed to plastic wrap or paper, these convenient markers emit low-level luminescence rather than radioactivity, making the Glogos II markers a safe, non-hazardous alternative to radioactive ink.