C18 EWP plates, Bond Elut
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7542601C 7540103C
VARI7542601CEA 6690 DKK
VARI7542601C VARI7540103C
C18 EWP plates, Bond Elut
Mikroplader Solid Phase Ekstraktions plader
Bond Elut C18 EWP uses a special silica with wide pores. This is endcapped.
  • No exclusion of large molecules
  • Good for desalting proteins
  • Successful separation of proteins, peptides or nucleotides
This is based upon standard particles size silica but with 500 Å pores to allow more efficient extraction of large molecules (>15000 MW), which are typically excluded from standard porosity silica phases.