Erlenmeyer flasks, narrow or wide neck, Super Duty, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
212172404 212275408 212173606 212174405 212175401 212273604 212274403 212171408 212171708 212176303 212177308 212272402
214-0212EA 181 DKK
214-0212 214-0221 214-0213 214-0214 214-0215 214-0219 214-0220 214-0210 214-0211 214-0216 214-0217 214-0218
Erlenmeyer flasks, narrow or wide neck, Super Duty, DURAN®
Kolber Erlenmeyer-kolber
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With easy to read graduations and a white labelling area. Higher mechanical stability compared to standard Erlenmeyer flasks due to increased wall thickness, plus reinforced sturdy rim and base. Even distribution of wall thickness. It is recommended to heat Super Duty Erlenmeyer flasks slowly and evenly, to prevent thermal stresses in the glass.
With printed Retrace Code (batch identification).