GC columns, J&W CP-Wax 58 FFAP CB
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CP7727 CP7747 CP7757 CP7767 CP7777 CP7717 CP7797
VARICP7727EA 16070 DKK
VARICP7727 VARICP7747 VARICP7757 VARICP7767 VARICP7777 VARICP7717 VARICP7797
GC columns, J&W CP-Wax 58 FFAP CB
Kromatografikolonner
Agilent JW CP-Wax 58 FFAP CB column is a nitroterephthalic acid-modified, chemically bonded polyethylene glycol phase.
  • Highest polarity bonded wax column for analysing polar compounds
CP-Wax 58 FFAP is designed for acidic compounds such as phenols and free fatty acids, both derivatised and underivatised.