Vacin and Went modified orchid basal salt mixture
Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PMV505A-10X1L PMV505B-10L PMV505C-50L
APOSPMV505A-10X1LEA 275 DKK
APOSPMV505A-10X1L APOSPMV505B-10L APOSPMV505C-50L
Vacin and Went modified orchid basal salt mixture
Celledyrkningsmedier
Contains an equivalent iron molar concentration of ferrous sulfate in place of ferric tartrate.
  • Plant tissue culture tested