SPE cartridges, Bond Elut™ Plexa PAX
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
12107301 12107603
VARI12107301EA 4410 DKK
VARI12107301 VARI12107603
SPE cartridges, Bond Elut™ Plexa PAX
Kromatografikolonner
Bond Elut™ plexa PAX non-polar/anion-exchange polymeric SPE offers excellent clean-up and reproducibility, and is suitable for polar and non-polar acidic analytes.
  • Mixed mode nonpolar polymeric anion exchanger offers high level of analyte selectivity
  • Exclusion of endogenous interferences offers superior cleanliness and minimises ion suppression
  • Simple, single method for ease-of-use reduces method development time
Existing polymeric anion-exchange sorbents can exhibit a broad range of ion exchange capacity from batch to batch. This leads to nonreproducible methods and compromised data. Plexa PAX particles are functionalised using a proprietary process, which allows anion exchange loadings to be controlled with a high degree of reproducibility, allowing a more robust performance across the lifetime of your compound study or method.