Brilliant II probe MM with ROX
Leverandør: AGILENT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
600805 600806
AGLS600805EA 4075 DKK
AGLS600805 AGLS600806
Brilliant II probe MM with ROX
Nukleinsyrereagenser qPCR/RT-qPCR Enzymes and Kits
Brilliant II QPCR and QRT-PCR reagents were developed to maintain the high quality of original Brilliant product line, but with a significant improvement in sensitivity of detection, ensuring reproducible quantification even at low target concentrations.
  • Formulations available with low and high concentrations of ROX reference dye
  • Improved performance with earlier Ct detection across a wide dynamic range
  • Increased sensitivity of detection to low copy numbers
With the addition of ROX reference dye to our master mix, the reaction setup process is streamlined.
Advarsel: For research use only. Not for use in diagnostic procedures.