VWR® Releasable Cable Ties, Polyamide 6,6
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
129-1623EA 326 DKK
129-1623 129-1624 129-1625 129-1626 129-1627
VWR® Releasable Cable Ties, Polyamide 6,6
Sikringsstrop
An effective, long lasting closure system. Releasable cable ties are intended for use when reopening is required.
  • High resistance to bases, oils, greases and oil derivatives
  • Limited resistance to acids
  • Not resistant to phenols or chloride solvents
  • Operating temperature: −40 to +85 °C
  • Installation temperature: −10 to +60 °C
  • Melting point: +260 °C
  • Flammability class V2 acc- a UL 94
Versatile releasable cable ties are available in several different sizes. The positive locking features of these ties enable the user to make precise tension adjustments. Easy to use in a wide range of applications, they provide a secure fastening solution.