ISIS supplies for 7500
Leverandør: AGILENT HEWLETT PACKARD
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5064-8016 5064-8017 5064-8018 5064-8026 5064-8021 5064-8022 5064-8020 G3138-65121 5064-8027 5064-8019 5064-8029 5064-8032 5064-8033 G3138-65117 G3138-65118 G3138-65119 G3138-65122 G3138-65125 G3138-65158
HEWL5064-8016EA 918 DKK
HEWL5064-8016 HEWL5064-8017 HEWL5064-8018 HEWL5064-8026 HEWL5064-8021 HEWL5064-8022 HEWL5064-8020 HEWLG3138-65121 HEWL5064-8027 HEWL5064-8019 HEWL5064-8029 HEWL5064-8032 HEWL5064-8033 HEWLG3138-65117 HEWLG3138-65118 HEWLG3138-65119 HEWLG3138-65122 HEWLG3138-65125 HEWLG3138-65158
ISIS supplies for 7500
Kromatografi forsyninger Kromatografi reservedele
High quality supplies and components for the Integrated Sample Introduction System (ISIS) for use with the 7500 ICP-MS range.
  • ETFE sample tubing with a 0,8 mm Int.Ø and 1,6 mm Ext.Ø is also available along with PTFE sample tubing with a 2 mm Int.Ø and 3 mm Int.Ø
  • Alternatively, PFA sample tubing with 0,3 mm Int.Ø and 1,6 mm Ext.Ø can be provided
  • A variety of joints are available in tee, cross or union form
  • PTFE nuts and front and back ferrules for either 3 mm or 1,6 mm Ext.Ø can be provided
  • Spiral tubing, peristaltic pump tubing adapters and tubing clamps, in small or large models, may be purchased
  • Wrenches for valve maintenance and tags for tubing identification may also be provided
  • A range of standard accessory parts for high sample throughput, auto dilution or discrete sampling are provided
Agilent's unique ISIS enhances the functionality and flexibility of the Agilent 7500 Series ICP-MS. ISIS features high precision peristaltic pumps and an optional switching valve to allow complete flexibility of configuration. The ISIS hardware is built into the 7500 chassis and close-coupled to increase productivity through high-speed sample uptake and rapid washout. To enhance the capabilities that the system offers and maintain performance even after heavy use, Agilent provides a range of replacement components and other supplies.