Ichihashi new phalaenopsis (NP) medium
Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PMI365A-10X1L PMI365B-10L PMI365C-50L
APOSPMI365A-10X1LEA 0 DKK
APOSPMI365A-10X1L APOSPMI365B-10L APOSPMI365C-50L
Ichihashi new phalaenopsis (NP) medium
Celledyrkningsmedier
Contains the components as described by Ichihashi (1992); modified to contain 82,0 mg/L NH₄NO₃.