Yeast extract
Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIY1333-1KG BIY1333-250G BIY1333-500G
APOSBIY1333-1KGEA 2650 DKK
APOSBIY1333-1KG APOSBIY1333-250G APOSBIY1333-500G
Yeast extract
Mikrobiologiske medier Dehydreret Mikrobiologisk medie
A dried yeast autolysate with a high content of amino nitrogen and water soluble B-complex vitamins.
  • MFCD00132599