Ethanol denatured, EMSURE® for analysis (denatured with 1% MEK, 1% IPA and 10 ppm Bitrex®)
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Ethyl alcohol denatured
1.03771.2500 1.03771.9180
1.03771.2500EA 656.25 DKK
1.03771.2500 1.03771.9180
Ethanol denatured, EMSURE® for analysis (denatured with 1% MEK, 1% IPA and 10 ppm Bitrex®)
Ethanol denatured
Bitrex = Denatonium benzoate
Formel: H₃CCH₂OH
Molekylvægt: 46.07 g/mol
Massefylde: 0.79 g/cm³(20 °C)
Flammepunkt: 12 °C
MDL Number: MFCD00003568
CAS-nummer: 64-17-5
UN nummer: 1170
ADR: 3 II